Order Entry
Netherlands
Orders LinkContactUsLinkComponent
1,5-Pentanedithiol 96%

1,5-Pentanedithiol 96%

Glass bottle for solids, 25 g
Catalog # B25665.14
Irritant
1,5-Pentanedithiol 96%
Catalog # B25665.14
CAS Number:  928-98-3
1,5-Pentanedithiol 96%
1,5-Pentanedithiol 96%

Glass bottle for solids, 25 g

CAS Number:  928-98-3
Price
82,50 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 96% CAS No.: 928-98-3 Molecular Formula: HS(CH2)5SH Formula Weight: 136.28 Flash Point: 95°(203°F) Refractive Index: 1.5194 Odour: Unpleasant Melting Point: -72° Boiling Point: 216-218° Beilstein Reference: 1732335 UN No.: 3334 MDL No.: MFCD00004908

Synonyms:  1,5-Dimercaptopentane
  • CAS Number:928-98-3
  • Formula:C₅H₁₂S₂
  • MDL Number:MFCD00004908
  • Molecular Weight:136.28 g/mol
  • Boiling Pt:216…218 °C
  • Melting Pt:–72 °C
  • Density:1.016
  • Flash Pt:95 °C (203 °F)
  • Storage Temperature:Ambient
  • EINECS:213-194-9
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.