Order Entry
Netherlands
Orders LinkContactUsLinkComponent
m-Xylene ≥99%

m-Xylene ≥99%

Glass bottle, 2.500 mL
Catalogus # L03788.0F
Ontvlambaar
Irriterend
Leverancier:   Thermo Fisher Scientific
m-Xylene ≥99%
Catalogus # L03788.0F
CAS-nummer:  108-38-3
m-Xylene ≥99%
m-Xylene ≥99%

Glass bottle, 2.500 mL

CAS-nummer:  108-38-3
Price
160,00 €
Log in voor beschikbaarheid
1 * 2.500 mL
Product Details & Documents

Purity: 99% CAS No.: 108-38-3 Molecular Formula: C8H10 Formula Weight: 106.17 Flash Point: 25°(77°F) Refractive Index: 1.4970 Odour: Mild Melting Point: -47° Boiling Point: 138-139° Beilstein Reference: 605441 Merck Reference: 14,10081 UN No.: 1307 MDL No.: MFCD00008536

Synoniemen:  1,3-Dimethylbenzene
  • CAS-nummer:108-38-3
  • Formule:C₈H₁₀
  • MDL-nummer:MFCD00008536
  • Molecular Weight:106.17 g/mol
  • Merck index:14,10081
  • Kokend Pt:138…139 °C
  • Smelten Pt:–47 °C
  • Dichtheid:0.866
  • Flits Pt:25 °C (77 °F)
  • Temperatuur bij opslag:Ambient
  • EINECS:203-576-3
  • UN:1307
  • ADR:3,II
Specificaties

Specificaties

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL

Specificaties