Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Basic cleanroom benches
Basic cleanroom benches
Basic cleanroom bench, 1200×350×450 mm
Catalog # 134-0499
Supplier:   Palbam
Basic cleanroom benches
Catalog # 134-0499
Supplier:  Palbam
Basic cleanroom benches
Basic cleanroom benches
Basic cleanroom bench, 1200×350×450 mm
Price
1 330,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Cleanroom benches in stainless steel 304 with electropolished finish.
  • Rounded edges
  • Height adjustable legs
  • Leg covers (covers unsightly bolt connections to floor)
  • No snag points
  • Ultra-clean
Delivery information: Typically available from stock for direct shipment from the supplier.
Specifications
  • Description:
    Basic cleanroom bench
  • Seq:
    04
  • W×D×H:
    1200×350×450 mm