Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Additional Top and Bottom Shelves
Top shelf, solid, for cleanroom heavy duty tables, W×D×H: 900×300×700 mm, fabricated from stainless steel 304 with electropolished finish
Catalogus # 132-0378
Leverancier:   Palbam
Additional Top and Bottom Shelves
Catalogus # 132-0378
Leverancier:  Palbam
Additional Top and Bottom Shelves
Additional Top and Bottom Shelves
Top shelf, solid, for cleanroom heavy duty tables, W×D×H: 900×300×700 mm, fabricated from stainless steel 304 with electropolished finish
Price
1 200,00 €
Log in voor beschikbaarheid
1 * 1 ST
Specificaties
  • Description:
    Top shelf, solid, for cleanroom heavy duty tables, W×D×H: 900×300×700 mm, fabricated from stainless steel 304 with electropolished finish
  • Seq:
    20