Order Entry
Netherlands
ContactUsLinkComponent
Anticon* Sterile 100 Cleanroom Wipers. For pharmaceutical, biotech, medical device manufacturing. 100% continuous filament polyester double-knit. Laundered cut edges. Made in ISO Class 1 facility. Sterilized to 1e-6 Sterility Assurance Level. Certif 1 * 100 items
Catalog # 115-0438
Supplier:  CONTEC
Anticon* Sterile 100 Cleanroom Wipers. For pharmaceutical, biotech, medical device manufacturing. 100% continuous filament polyester double-knit. Laundered cut edges. Made in ISO Class 1 facility. Sterilized to 1e-6 Sterility Assurance Level. Certif 1 * 100 items
Catalog # 115-0438
Supplier:  CONTEC
Frequently Bought Together