Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Hypoxanthine ≥99%

Hypoxanthine ≥99%

Glass bottle for solids, 25 g
Catalog # A11481.14
Hypoxanthine ≥99%
Catalog # A11481.14
CAS Number:  68-94-0
Hypoxanthine ≥99%
Hypoxanthine ≥99%

Glass bottle for solids, 25 g

CAS Number:  68-94-0
Price
81,80 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 99% CAS No.: 68-94-0 Molecular Formula: C5H4N4O Formula Weight: 136.11 Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 5811 Merck Reference: 14,4869 MDL No.: MFCD00005725

Synonyms:  1H-Purin-6-one, 1,9-Dihydro-6H-purin-6-one, Purin-6(1H)-one, 6-Purinone
  • CAS Number:68-94-0
  • Formula:C₅H₄N₄O
  • MDL Number:MFCD00005725
  • Molecular Weight:136.11 g/mol
  • Merck Index:14,4869
  • Melting Pt:>300 °C
  • EINECS:200-697-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.