Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Lamivudine, Reference Standards

Lamivudine, Reference Standards

Lamivudine, USP reference standard, Vial, 200 mg
Catalog # 1356836.
Supplier:   USP
Lamivudine, Reference Standards
Catalog # 1356836.
Supplier:  USP
CAS Number:  
Lamivudine, Reference Standards

Lamivudine, USP reference standard, Vial, 200 mg

CAS Number:  
Price
392,00 €
Log in for availability
1 * 200 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Lamivudine Resolution Mixture B: This is a mixture of lamivudine (about 94%) and lamivudine diastereomer (about 6%)

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • Description:
    Lamivudine
  • Cat. No.:
    1356836.
  • Formula:
    C₈H₁₁N₃O₃S
  • Description:
    Lamivudine
  • CAS No.:
    134678-17-4

Specifications