Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Histosette® II, M493
Biopsy processing/embedding cassettes, Histosette® II, M493
Salmon
Catalog # 720-0924
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Histosette® II, M493
Catalog # 720-0924
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Histosette® II, M493
Biopsy processing/embedding cassettes, Histosette® II, M493
Salmon
Price
292,00 €
Log in for availability
1 * 1.500 ST
Product Details & Documents
Acetal polymer.
  • No need for biopsy pads
  • Resistant to most commonly used histological solutions
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems
Specifications
  • Colour:
    Salmon