Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Paraformaldehyde

Paraformaldehyde

5 kg
Catalog # ROTH0335.4
Flammable
Health
Irritant
Marketsource
Supplier:   Roth Carl
Paraformaldehyde
Catalog # ROTH0335.4
Supplier:  Roth Carl
Marketsource
CAS Number:  30525-89-4
Paraformaldehyde

5 kg

CAS Number:  30525-89-4
Price
Inquire For Price
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Polyoxymethylene
  • CAS Number:30525-89-4
  • Formula:(CH₂O)n
  • MDL Number:MFCD00133991
  • Merck Index:13,07096
  • Boiling Pt:135…136 °C (13 mmHg)
  • Melting Pt:123 °C
  • Density:1.46 g/cm³ (20 °C)
  • Flash Pt:70 °C
  • Storage Temperature:Ambient
  • UN:2213
  • ADR:4.1,III
Specifications

Specifications

  • Pk:
    5 kg

Specifications