Tween® 20 (Polysorbate)
Glass bottle for solids, 1 kg
Catalog # ROTH9127.2
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Polyoxyethylene sorbitan monolaurate
- CAS Number:9005-64-5
- Formula:C₅₈H₁₁₄O₂₆
- MDL Number:MFCD00165986
- Molecular Weight:1227.53 g/mol
- Density:1.1 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:500-018-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



