Diethyl ether
5 L
Catalog # ROTH5920.3
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
- CAS Number:60-29-7
- Formula:(CH₃CH₂)₂O
- MDL Number:MFCD00011646
- Molecular Weight:74.12 g/mol
- Merck Index:12,03852
- Boiling Pt:35 °C (1013 hPa)
- Melting Pt:–123 °C
- Density:0.7135 g/cm³ (20 °C)
- Flash Pt:–40 °C
- Storage Temperature:Ambient
- EINECS:200-467-2
- UN:1155
- ADR:3,I
- REACH:01-2119535785-29
Specifications
Specifications
- Pk:5 L



