Trypsin
Glass bottle for solids, 100 mg
Catalog # ROTH5455.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Documents
ROTH5455.1_TDS_EN
Datasheet
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Cat. No.:ROTH5455.1
- Storage conditions:Refrigerator
- Purity:1× crystallized
- MDL No.:MFCD00082094
- Enzyme name:Trypsin
- CAS No.:9002-07-7
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



