Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
100 ml, 19/26, 0,100 ± ml
Catalog # 612-5872
Supplier:   HIRSCHMANN
Volumetric flasks, class A, blue markings
Catalog # 612-5872
Supplier:  HIRSCHMANN
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
100 ml, 19/26, 0,100 ± ml
Price
72,40 €
Log in for availability
1 * 2 ST
Product Details & Documents
DURAN®, borosilicate glass, with PE stopper, conformity certified (DE-M marking).
  • Blue enamel indelible markings resistant to acids and alkalis
  • Certificate of conformity with dated batch identification available via website (www.hirschmannlab.de)
* In addition to DIN.
**Wide neck
Certifications: DIN EN ISO 1042
Specifications
  • NS:
    19/26
  • Capacity:
    100 ml
  • Seq:
    13
  • Tolerance:
    0,100 ± ml