Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Calibration weights, class F1

Calibration weights, class F1

5 g, 0,16 ± mg, Cylindrical test weight
Catalog # SARBYCW3538-00
Supplier:   Sartorius Balances
Calibration weights, class F1
Catalog # SARBYCW3538-00
Supplier:  Sartorius Balances
Calibration weights, class F1
Calibration weights, class F1

5 g, 0,16 ± mg, Cylindrical test weight

Price
72,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

Cylindrical weights which are made up of special stainless steel with a density of 7,950 kg/m³.

Certifications: Comply in all respects with the requirements on shape, material and markings specified by the OIML R111-1 International Recommendation.

Delivery information: Cylindrical weights are supplied in high-quality plastic cases that are suitable for use in cleanrooms.

Specifications
  • Weight:
    5 g
  • Seq:
    03
  • Tolerance:
    0,16 ± mg
  • Type:
    Cylindrical test weight