Order Entry
Netherlands
ContactUsLinkComponent
Thermometer, refill, cylinder, mercury filled, blue reflecting, measuring range:0:100/1C, colourless, enclosed scale, capillary le n s front, dimension:250x17mm, cork glass top finish. Until 10th October 2017 it is still allowed to use mercury ther 1 * 1 items
Catalog # AMART108294
Supplier:  Amarell
Thermometer, refill, cylinder, mercury filled, blue reflecting, measuring range:0:100/1C, colourless, enclosed scale, capillary le n s front, dimension:250x17mm, cork glass top finish. Until 10th October 2017 it is still allowed to use mercury ther 1 * 1 items
Catalog # AMART108294
Supplier:  Amarell
Frequently Bought Together