Order Entry
Netherlands
ContactUsLinkComponent
Thermometer, colorimeter, mercury filled, Berthelot-Mahler, measuring range:15:25/0.01C, with glass saddle and metal cap, milk gla s s scale, capillary round, colourless, upper part: 580x14-15mm, lower part :210x10-13mm, immersion:200mm, suitable f 1 * 1 items
Catalog # AMARK164006
Supplier:  Amarell
Thermometer, colorimeter, mercury filled, Berthelot-Mahler, measuring range:15:25/0.01C, with glass saddle and metal cap, milk gla s s scale, capillary round, colourless, upper part: 580x14-15mm, lower part :210x10-13mm, immersion:200mm, suitable f 1 * 1 items
Catalog # AMARK164006
Supplier:  Amarell
Frequently Bought Together