Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Dispenser tips, ritips®

Dispenser tips, ritips®

Bioclean®, 50 ml, +, Ritter ripette®, Eppendorf® Multipette 4780, 25/pack
Catalogus # BZYM742116
Marketsource
Leverancier:   BIOZYM
Dispenser tips, ritips®
Catalogus # BZYM742116
Leverancier:  BIOZYM
Marketsource
Dispenser tips, ritips®

Bioclean®, 50 ml, +, Ritter ripette®, Eppendorf® Multipette 4780, 25/pack

Price
Inquire For Price
Log in for availability
1 * 25 ST
Product Details & Documents

These dispenser tips are compatible with Ritter ripette®, Eppendorf Multipette® 4780 and many other dispensing systems.

Bioclean® tips are sterile and free from DNase, RNase and pyrogens.

Delivery information: The 25 ml and 50 ml tips are supplied with adapter.

Specifications
  • Packed:
    25/pack
  • Sterile:
    +
  • Volume:
    50 ml
  • Tip style:
    Bioclean®
  • For use with:
    Ritter ripette®, Eppendorf® Multipette 4780
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.