Order Entry
Netherlands
Orders LinkContactUsLinkComponent
E.Z.N.A.® Plasmid Mini kit I

E.Z.N.A.® Plasmid Mini kit I

E.Z.N.A.® Plasmid Mini Kit I (V-Spin column), 200 Tests
Catalogus # D6943-02
Leverancier:   OMEGA BIO-TEK
E.Z.N.A.® Plasmid Mini kit I
Catalogus # D6943-02
Leverancier:  OMEGA BIO-TEK
CAS-nummer:  
E.Z.N.A.® Plasmid Mini kit I
E.Z.N.A.® Plasmid Mini kit I

E.Z.N.A.® Plasmid Mini Kit I (V-Spin column), 200 Tests

CAS-nummer:  
Price
249,00 €
Log in voor beschikbaarheid
1 * 1 EA
Product Details & Documents

The E.Z.N.A.® Plasmid Mini Kit I is designed to isolate up to 30 µg of high-quality plasmid DNA from 1 to 5 ml bacterial cultures in 30 minutes or less. Plasmid DNA purification is simplified with HiBind® Mini Column technology into three quick steps: Bind, wash, and elute. Purified plasmid DNA is immediately ready for a wide variety of downstream applications such as routine screening, restriction enzyme digestion, and manual/automated fluorescent DNA sequencing. E.Z.N.A.® Plasmid Mini Kit I is available in either V-spin or Q-spin column format. V-spin column formats have an attached lid while Q-spin columns do not. Both versions can be used for centrifugation or vacuum protocols.

  • Purification of plasmid DNA in 30 minutes or less
  • No phenol/chloroform extractions
  • DNA suitable for downstream applications
Specificaties

Specificaties

  • Pk:
    200 Tests
  • Description:
    E.Z.N.A.® Plasmid Mini Kit I (V-Spin column)
  • Cat. No.:
    D6943-02
  • Prep size:
    Mini
  • Refine target molecule:
    Plasmid
  • Target molecule:
    Plasmid
  • Method / Format:
    Column (silica-based)
  • Description:
    E.Z.N.A.® Plasmid Mini Kit I (V-Spin column)
  • Sample type:
    Bacteria
  • Sample size:
    1 - 5 ml

Specificaties

Aanbevelingen

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.