Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Dnase
Dnase
50 Tests
Catalog # E1091
Supplier:   OMEGA BIO-TEK
Dnase
Catalog # E1091
Supplier:  OMEGA BIO-TEK
CAS Number:  
Dnase
50 Tests
CAS Number:  
Price
125,00 €
Log in for availability
1 * 1 EA
Product Details & Documents
For DNase digestion during RNA purification. The RNase-free DNase set provides a rapid and efficient on-column digestion of DNA during RNA purification from a wide variety of sample sources using E.Z.N.A.® RNA kits. The DNases can be efficiently removed in subsequent steps. Generally, DNase digestion is not required for RNA purified with E.Z.N.A.® RNA kits since the silica-membrance, spin-column technology efficiently removes the majority of the DNA without DNase treatment. However, complete DNA removal may be necessary for certain RNA applications that are sensitive to very small amounts of DNA.
  • RNase-free
Specifications
Specifications
  • Pk:
    50 Tests
  • Cat. No.:
    E1091
  • Enzyme name:
    Dnase