Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Calibration weights, class F1

1 kg, 5,0 ± mg, Cylindrical test weight
Catalog # SARBYCW6138-02
Supplier:   Sartorius Balances
Calibration weights, class F1
Catalog # SARBYCW6138-02
Supplier:  Sartorius Balances
Calibration weights, class F1
Calibration weights, class F1

1 kg, 5,0 ± mg, Cylindrical test weight

Price
369,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Cylindrical weights which are made up of special stainless steel with a density of 7,950 kg/m³.

 : Comply in all respects with the requirements on shape, material and markings specified by the OIML R111-1 International Recommendation.

 : Cylindrical weights are supplied in high-quality plastic cases that are suitable for use in cleanrooms.

Specifications
  • Weight:
    1 kg
  • Seq:
    21
  • Tolerance:
    5,0 ± mg
  • Type:
    Cylindrical test weight