Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

L(+)-Glutamine, Millipore®

Plastic drum, 10 kg
Catalog # 1.00289.9010
Supplier:   Merck
L(+)-Glutamine, Millipore®
Catalog # 1.00289.9010
Supplier:  Merck
CAS Number:  56-85-9
L(+)-Glutamine, Millipore®
L(+)-Glutamine, Millipore®

Plastic drum, 10 kg

CAS Number:  56-85-9
Price
Inquire For Price
Log in for availability
1 * 10 kg
Product Details & Documents
Synonyms:  Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
  • CAS Number:56-85-9
  • Formula:C₅H₁₀N₂O₃
  • MDL Number:MFCD00008044
  • Molecular Weight:146.15 g/mol
  • Merck Index:13,04484
  • Boiling Pt:355 °C (1013 hPa)
  • Melting Pt:185…186 °C
  • Density:1.5326 g/cm³ (130 °C)
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:200-292-1
Specifications

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    10 kg

Specifications