Slide containers, SlideRite®
Transport containers for slides, SlideRite I, 1
Catalog # 720-2326
Supplier: CELLPATH
Jump to:
Product Details & Documents
Slide holders designed for use with one or two slides.
- Slide- and a click-closed lid
- Slide stays in the container to apply the swab
Suitable for shipping, storing, or staining slides.
Specifications
- No. of slides:1
- Description:Transport containers for slides, SlideRite I
- Cat. No.:720-2326