Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Borane trimethylamine complex 97%
Glass bottle for solids, 25 g
  L14994.14
Borane trimethylamine complex 97%
  L14994.14
 :  75-22-9
Borane trimethylamine complex 97%
Borane trimethylamine complex 97%
Glass bottle for solids, 25 g
 :  75-22-9
85,40 €
1 * 25 g
Purity: 97% CAS No.: 75-22-9 Molecular Formula: (CH3)3N.BH3 Formula Weight: 72.95 Flash Point: 64°(147°F) Melting Point: 93-95° Boiling Point: 172° Beilstein Reference: 3905060 UN No.: 1325 MDL No.: MFCD00012420
  •  :75-22-9
  •  :C₃H₁₂BN
  •  :MFCD00012420
  •  :72.95 g/mol
  •  :172 °C
  •  :93…95 °C
  •  :0.81
  •  :64 °C (147 °F)
  •  :Ambient
  •  :200-850-4
  •  :1325
  •  :4.1,II
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g