Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Borane trimethylamine complex 97%

Borane trimethylamine complex 97%

Glass bottle for solids, 5 g
Catalogus # L14994.06
Flammable
Irritant
Leverancier:   Thermo Fisher Scientific
Borane trimethylamine complex 97%
Catalogus # L14994.06
CAS-nummer:  75-22-9
Borane trimethylamine complex 97%
Borane trimethylamine complex 97%

Glass bottle for solids, 5 g

CAS-nummer:  75-22-9
Price
30,20 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 97% CAS No.: 75-22-9 Molecular Formula: (CH3)3N.BH3 Formula Weight: 72.95 Flash Point: 64°(147°F) Melting Point: 93-95° Boiling Point: 172° Beilstein Reference: 3905060 UN No.: 1325 MDL No.: MFCD00012420

  • CAS Number:75-22-9
  • Formula:C₃H₁₂BN
  • MDL Number:MFCD00012420
  • Molecular Weight:72.95 g/mol
  • Boiling Pt:172 °C
  • Melting Pt:93…95 °C
  • Density:0.81
  • Flash Pt:64 °C (147 °F)
  • Storage Temperature:Ambient
  • EINECS:200-850-4
  • UN:1325
  • ADR:4.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.