Order Entry
Netherlands
ContactUsLinkComponent
RFM830 DIGITAL REFRACTOMETER 1.32-1.58/0.0001RI, 0-100/0.1 BRIX, PELTIER TEMPERATURE CONTROL 1 * 1 items
Catalog # 635-0365
Supplier:  BELLINGHAM STANLEY
RFM830 DIGITAL REFRACTOMETER 1.32-1.58/0.0001RI, 0-100/0.1 BRIX, PELTIER TEMPERATURE CONTROL 1 * 1 items
Catalog # 635-0365
Supplier:  BELLINGHAM STANLEY
Frequently Bought Together