N-[(4-chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridin-5-yl)carbonyl]-N'-[3,5-di(trifluoromethyl)phenyl]urea, TECH 1 * 250 mg
Catalog # MAYBKM03175.250
Supplier: Thermo Scientific
N-[(4-chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridin-5-yl)carbonyl]-N'-[3,5-di(trifluoromethyl)phenyl]urea, TECH 1 * 250 mg
Catalog # MAYBKM03175.250
Supplier: Thermo Scientific
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck