Melatonin 99%
Glass bottle for solids, 250 mg
Leverancier: Thermo Scientific
Melatonin 99%
Catalogus # MAYBAC12536.250
Leverancier: Thermo Scientific
Marketsource
CAS-nummer: 73-31-4
Jump to:
Product Details & Documents
Synonyms: Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
- CAS Number:73-31-4
- Formula:C₁₃H₁₆N₂O₂
- MDL Number:MFCD00005655
- Molecular Weight:232.28 g/mol
- Boiling Pt:512.8 °C
- Melting Pt:118 °C
- Flash Pt:264 °C
- Storage Temperature:Ambient
- EINECS:200-797-7
Specifications
Specification Test Results
- Appearance:White to cream to yellow Powder
- Infrared spectrum:Conforms
- Melting point:116 to 120 °C
- Loss on drying:≤0.5 % (1 g, 105 °C)
- Heavy metals:≤20 ppm
- HPLC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



