1,2,3,4-TETRAHYDRO-1-NAPHTHONITRILE 1 * 100 mg
Catalog # PROOMM0088.03
Supplier: LGC Standards PROMOCHEM
1,2,3,4-TETRAHYDRO-1-NAPHTHONITRILE 1 * 100 mg
Catalog # PROOMM0088.03
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Iso-Hexane mixture of isomers ≥97.0% (mixture of isomers), HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
N-Ethyldiisopropylamine ≥99.5% for peptide synthesis
Supplier: VWR Chemicals
Acetonitrile : water : trifluoroacetic acid (50 : 47,5 : 2,5), HiPerSolv CHROMANORM®
Supplier: VWR Chemicals
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Cyclohexane, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Benzyl alcohol 99%, pure
Supplier: Thermo Fisher Scientific
1-Butylboronic acid, Supelco®
Supplier: Merck
2-Propanol ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals