Handheld refractometers
40 - 82 °Brix, 0,5 °Brix
Catalog # 635-0312
Supplier: EUROMEX
Jump to:
Product Details & Documents
Single dual or triple scales, analogue, handheld refractometers.
- Automated temperature compensation (ATC)
- Calibrated at 20 °C except for RF.5190
- Requires only a few drops of liquid
Specifications
- Range:40 - 82 °Brix
- Seq:05
- Precision:0,5 °Brix