Order Entry
Netherlands
ContactUsLinkComponent
Anti-Laminin a2 Antibody, clone 5H2, Azide Free Anti-Laminin a2 Antibody, clone 5H2, Azide Free detects level of Laminin a2 & has been published & validated for use in ELISA, IH, IP & WB. Storage Conditions: Maintain between 2 and 8°C for up to 6 mon 1 * 100 µG
Catalog # CHMCMAB1922Z
Supplier:  Chemicon
Anti-Laminin a2 Antibody, clone 5H2, Azide Free Anti-Laminin a2 Antibody, clone 5H2, Azide Free detects level of Laminin a2 & has been published & validated for use in ELISA, IH, IP & WB. Storage Conditions: Maintain between 2 and 8°C for up to 6 mon 1 * 100 µG
Catalog # CHMCMAB1922Z
Supplier:  Chemicon
Frequently Bought Together