Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

HPLC columns, ZORBAX® Eclipse XDB

Rapid resolution HT cartridge, Guard cartridge, Eclipse XDB-C18, 1,8 µm, 4,60 mm, 30 mm
Catalog # HEWL923975-902
Supplier:   Agilent
HPLC columns, ZORBAX® Eclipse XDB
Catalog # HEWL923975-902
Supplier:  Agilent
HPLC columns, ZORBAX® Eclipse XDB
HPLC columns, ZORBAX® Eclipse XDB

Rapid resolution HT cartridge, Guard cartridge, Eclipse XDB-C18, 1,8 µm, 4,60 mm, 30 mm

Price
593,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Good peak shape for basic, acidic and neutral compounds.

  • High performance over a wide pH range (2 to 9)
  • Long lifetime with extra dens bonding and double endcapping
Specifications
  • Column Format:
    Guard cartridge
  • Ø int.:
    4,60 mm
  • Particle Size:
    1,8 µm
  • Description:
    Rapid resolution HT cartridge
  • Length:
    30 mm
  • Phase:
    Eclipse XDB-C18