Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Double spatulas, micro, one conical end

Double spatulas, micro, one conical end

210 mm, 7 mm
Catalog # RSGA039.210
Supplier:   RSG
Double spatulas, micro, one conical end
Catalog # RSGA039.210
Supplier:  RSG
Double spatulas, micro, one conical end
Double spatulas, micro, one conical end

210 mm, 7 mm

Price
36,90 €
Log in for availability
1 * 5 ST
Product Details & Documents

Micro double spatulas with one conical end, 7 mm wide. Made of stainless steel.

Specifications
  • Length:
    210 mm
  • Blade width:
    7 mm