Order Entry
Netherlands
Orders LinkContactUsLinkComponent
TLC indicators

TLC indicators

Fluorescent indicator UV366 (blue), 100 g
Catalog # MANA816720.01
Supplier:   MACHEREY-NAGEL
TLC indicators
Catalog # MANA816720.01
Supplier:  MACHEREY-NAGEL
CAS Number:  
TLC indicators
TLC indicators

Fluorescent indicator UV366 (blue), 100 g

CAS Number:  
Price
79,80 €
Log in for availability
1 * 100 g
Product Details & Documents

Fluorescent indicators for thin layer chromatography.

  • UV indicators with efficient radiation for short-wave as well as long-wave UV ranges
  • Can be used for preparation of TLC plates

UV254: Manganese activated zinc silicate with absorption maximum at 254 nm; green fluorescence.

UV366: Inorganic fluorescent pigment with absorption maximum at 366 nm; blue fluorescence.

Documents

Specifications

Specifications

  • Pk:
    100 g
  • Description:
    Fluorescent indicator UV<sub>366</sub> (blue)

Specifications