Filter paper circles MN 704 15cm diameter, pack of 100. 1 * 100 items
Catalog # MANA451015
Supplier: MACHEREY-NAGEL
Filter paper circles MN 704 15cm diameter, pack of 100. 1 * 100 items
Catalog # MANA451015
Supplier: MACHEREY-NAGEL
Frequently Bought Together
VWR®, Glass Fibre Filters
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Weighing Paper, Grade 531
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Büchner Funnels
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Desiccator lids, DURAN®
Supplier: DWK Life Sciences
VWR®, Syringe Filters
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Glass microfibre filters without binder, grade GF/C, Whatman™
Supplier: Whatman products (Cytiva)
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection