Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

D-Cycloserine

Glass bottle for solids, 25 g
Catalog # A1943.0025
Irritant
Supplier:   PanReac AppliChem
D-Cycloserine
Catalog # A1943.0025
Supplier:  PanReac AppliChem
CAS Number:  68-41-7
D-Cycloserine

Glass bottle for solids, 25 g

CAS Number:  68-41-7
Price
1 220,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine
  • CAS Number:68-41-7
  • Formula:C₃H₆N₂O₂
  • MDL Number:MFCD00005353
  • Molecular Weight:102.09 g/mol
  • Storage Temperature:Freezer
  • EINECS:200-688-4

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.