D-(-)-Histidine
Glass bottle for solids, 100 g
Catalog # BACE4030626.0100
Marketsource
Supplier: BACHEM BIOCHEMICA
D-(-)-Histidine
Catalog # BACE4030626.0100
Supplier: BACHEM BIOCHEMICA
Marketsource
CAS Number: 351-50-8
Jump to:
Product Details & Documents
Synonyms: (R)-(-)-Histidine, D-3-Imidazol-4-ylalanine, (2R)-(-)-Histidine, H-D-His-OH
- CAS Number:351-50-8
- Formula:C₆H₉N₃O₂
- MDL Number:MFCD00065963
- Molecular Weight:155.16 g/mol
- Merck Index:12,04758
- Melting Pt:287 °C
- EINECS:206-513-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



