D(-)-Lactic acid
Glass bottle for solids, 250 mg
Catalogus # BACE4003254.0250
Marketsource
Leverancier: BACHEM BIOCHEMICA
D(-)-Lactic acid
Catalogus # BACE4003254.0250
Leverancier: BACHEM BIOCHEMICA
Marketsource
CAS-nummer: 10326-41-7
Jump to:
Product Details & Documents
Synonyms: (R)-2-Hydroxypropanoic acid, D-Lactic Acid, (R)-(-)-2-Hydroxypropionic acid, (R)-(-)-Lactic acid, (R)-(-)-2-Hydroxypropanoic acid
- CAS Number:10326-41-7
- Formula:C₃H₆O₃
- MDL Number:MFCD00068313
- Molecular Weight:90.08 g/mol
- Storage Temperature:Freezer
- EINECS:233-713-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



