(R)-(-)-2-Hydroxyisovaleric acid
Glass bottle for solids, 5 g
Catalog # BACE4001785.0005
Marketsource
Supplier: BACHEM BIOCHEMICA
(R)-(-)-2-Hydroxyisovaleric acid
Catalog # BACE4001785.0005
Supplier: BACHEM BIOCHEMICA
Marketsource
CAS Number: 17407-56-6
Jump to:
Product Details & Documents
Synonyms: D-α-Hydroxyisovaleric acid, 2-Hydroxyisovaleric-acid, (R)-2-Hydroxy-3-methylbutanoic acid, D-Valic acid, (2R)-2-Hydroxy-3-methylbutanoic acid
- CAS Number:17407-56-6
- Formula:C₅H₁₀O₃
- MDL Number:MFCD00066442
- Molecular Weight:118.13 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



