Order Entry
Netherlands
Orders LinkContactUsLinkComponent
(R)-(-)-2-Hydroxyisovaleric acid

(R)-(-)-2-Hydroxyisovaleric acid

Glass bottle for solids, 1 g
Catalog # BACE4001785.0001
Marketsource
Supplier:   BACHEM BIOCHEMICA
(R)-(-)-2-Hydroxyisovaleric acid
Catalog # BACE4001785.0001
Supplier:  BACHEM BIOCHEMICA
Marketsource
CAS Number:  17407-56-6
(R)-(-)-2-Hydroxyisovaleric acid

Glass bottle for solids, 1 g

CAS Number:  17407-56-6
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  D-α-Hydroxyisovaleric acid, 2-Hydroxyisovaleric-acid, (R)-2-Hydroxy-3-methylbutanoic acid, D-Valic acid, (2R)-2-Hydroxy-3-methylbutanoic acid
  • CAS Number:17407-56-6
  • Formula:C₅H₁₀O₃
  • MDL Number:MFCD00066442
  • Molecular Weight:118.13 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.