D(-)-Glutamine
Glass bottle for solids, 25 g
Catalog # BACE4007503.0025
Marketsource
Supplier: BACHEM BIOCHEMICA
D(-)-Glutamine
Catalog # BACE4007503.0025
Supplier: BACHEM BIOCHEMICA
Marketsource
CAS Number: 5959-95-5
Jump to:
Product Details & Documents
Synonyms: H-D-Gln-OH, (R)-2,5-Diamino-5-oxopentanoic acid, (R)-(-)-Glutamine
- CAS Number:5959-95-5
- Formula:C₅H₁₀N₂O₃
- MDL Number:MFCD00065607
- Molecular Weight:146.15 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



