D(-)-Glutamine
Glass bottle for solids, 1 g
Catalog # BACE4007503.0001
Marketsource
Supplier: BACHEM BIOCHEMICA
D(-)-Glutamine
Catalog # BACE4007503.0001
Supplier: BACHEM BIOCHEMICA
Marketsource
CAS Number: 5959-95-5
Jump to:
Product Details & Documents
Synonyms: H-D-Gln-OH, (R)-2,5-Diamino-5-oxopentanoic acid, (R)-(-)-Glutamine
- CAS Number:5959-95-5
- Formula:C₅H₁₀N₂O₃
- MDL Number:MFCD00065607
- Molecular Weight:146.15 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



