Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Calibration weights, class F1

Calibration weights, class F1

2 kg, 10,0 ± mg, Cylindrical test weight
Catalogus # 611-0899
Leverancier:   Sartorius Balances
Calibration weights, class F1
Catalogus # 611-0899
Leverancier:  Sartorius Balances
Calibration weights, class F1
Calibration weights, class F1

2 kg, 10,0 ± mg, Cylindrical test weight

Price
536,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

Cylindrical weights which are made up of special stainless steel with a density of 7,950 kg/m³.

Certifications: Comply in all respects with the requirements on shape, material and markings specified by the OIML R111-1 International Recommendation.

Delivery information: Cylindrical weights are supplied in high-quality plastic cases that are suitable for use in cleanrooms.

Specificaties
  • Weight:
    2 kg
  • Seq:
    22
  • Tolerance:
    10,0 ± mg
  • Type:
    Cylindrical test weight