HPLC columns, ZORBAX® Eclipse XDB
Hardware kit, Guard cartridge
Catalog # HEWL820555-901
Supplier: Agilent
Jump to:
Product Details & Documents
Good peak shape for basic, acidic and neutral compounds.
- High performance over a wide pH range (2 to 9)
- Long lifetime with extra dens bonding and double endcapping
Specifications
- Column Format:Guard cartridge
- Description:Hardware kit