Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Handheld refractometers
Handheld refractometers
0 - 80 °Brix, 0,5 °Brix
Catalog # 635-0081
Supplier:   EUROMEX
Handheld refractometers
Catalog # 635-0081
Supplier:  EUROMEX
Handheld refractometers
Handheld refractometers
0 - 80 °Brix, 0,5 °Brix
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Single dual or triple scales, analogue, handheld refractometers.
  • Automated temperature compensation (ATC)
  • Calibrated at 20 °C except for RF.5190
  • Requires only a few drops of liquid
Specifications
  • Range:
    0 - 80 °Brix
  • Seq:
    07
  • Precision:
    0,5 °Brix