Order Entry
Netherlands
ContactUsLinkComponent
Miq/cr3,module iq/current relais 3, for system 2020 xt and 184 xt, with 3 analog (0/4-20 ma) and 3 relay outputs (250 vac/5 a) 1 * 1 items
Catalog # WTWO480014
Supplier:  WTW ONLINE
Miq/cr3,module iq/current relais 3, for system 2020 xt and 184 xt, with 3 analog (0/4-20 ma) and 3 relay outputs (250 vac/5 a) 1 * 1 items
Catalog # WTWO480014
Supplier:  WTW ONLINE
Frequently Bought Together