Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Accessories for transilluminators

Replacement longwave (365 nm) UV tube, 25 W
Catalog # UVPA34-0060-01
Accessories for transilluminators
Catalog # UVPA34-0060-01
Accessories for transilluminators

Replacement longwave (365 nm) UV tube, 25 W

Price
53,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

 : For information about any replacement tubes that are not listed, please contact your local Avantor office.

Specifications
  • Description:
    Replacement longwave (365 nm) UV tube, 25 W
  • Seq:
    07