Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Accessories for transilluminators

Accessories for transilluminators

Replacement mid-range (254 nm) UV tube, 25 W
Catalog # UVPA34-0073-01
Accessories for transilluminators
Catalog # UVPA34-0073-01
Accessories for transilluminators

Replacement mid-range (254 nm) UV tube, 25 W

Price
67,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

Accessories information: For information about any replacement tubes that are not listed, please contact your local Avantor office.

Specifications
  • Description:
    Replacement mid-range (254 nm) UV tube, 25 W
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.