Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

3,3',5,5'-Tetramethylbenzidine (TMB), pure

Glass bottle for solids, 25 g
Catalog # BMOL22166-2
3,3',5,5'-Tetramethylbenzidine (TMB), pure
Catalog # BMOL22166-2
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB), pure

Glass bottle for solids, 25 g

CAS Number:  54827-17-7
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents

Use as water soluble, non-carcinogenic peroxidase substrate for ELISA application. Further applications: detection of cholesterol in blood serum, and blood in urine. Measure absorbance of the yellow reaction product at 450 nm.

Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Boiling Pt:402 °C (1013 hPa)
  • Melting Pt:166…170 °C
  • Storage Temperature:Ambient
  • EINECS:259-364-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications