Order Entry
Netherlands
ContactUsLinkComponent
Measuring cylinder 250 ml with glass base Weight and grading according to DIN12580, class B. 1 * 1 items
  827-4570
Measuring cylinder 250 ml with glass base Weight and grading according to DIN12580, class B. 1 * 1 items
  827-4570
Frequently Bought Together