Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Multiple socket outlet, six-Socket, economy

Multiple socket outlet, six-Socket, economy

Socket with switch and child protection, 6-way, 3 m
Catalog # 139-1330
Supplier:   BACHMANN ELEKTRO
Multiple socket outlet, six-Socket, economy
Catalog # 139-1330
Supplier:  BACHMANN ELEKTRO
Multiple socket outlet, six-Socket, economy
Multiple socket outlet, six-Socket, economy

Socket with switch and child protection, 6-way, 3 m

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Trailing socket with built-in, attractive and practical cable rewind.

  • Moulded, shock-proof plug with double earthing
  • Mounting flange for quick, easy attachment
  • Sockets offset by 35°

Colour: White

Specifications
  • Description:
    Socket with switch and child protection, 6-way
  • Length:
    3 m